Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FHL5
- Entrez Gene ID
-
9457
- Gene Name
- four and a half LIM domains 5
- Gene Aliases
-
1700027G07Rik, ACT, FHL-5, dJ393D12.2
- Chromosome Location
-
Chr.6: 96562525 - 96618626 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC011015.1 |
- |
1 - 1 |
453 |
71 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF673472.1 |
- |
1 |
446 |
|
Chr. 6 - 96563131 - 96563849 [+] on Build GRCh38 |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
UFM1 specific ligase 1 |
UFL1 |
KIAA0776, Maxer, NLBP, RCAD |
-7.9kb
|
Sense |
Chr. - 96521826 - 96555276 on Build GRCh38 |
Downstream |
G protein-coupled receptor 63 |
GPR63 |
PSP24(beta), PSP24B |
230.0kb
|
Anti-sense |
Chr. - 96794122 - 96837477 on Build GRCh38 |
View Full Product Details