Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIGL
Entrez Gene ID
9487
Gene Name
phosphatidylinositol glycan anchor biosynthesis class L
Gene Aliases
CHIME
Chromosome Location
Chr.17: 16217195 - 16351800 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017025349.1 XP_016880838.1 9 - 9 5631 105
XM_017025350.1 XP_016880839.1 9 - 9 5626 105
XM_017025352.1 XP_016880841.1 10 - 10 4799 105
XM_017025353.1 XP_016880842.1 9 - 9 4651 105
XM_017025354.1 XP_016880843.1 9 - 9 4767 105
XM_017025355.1 XP_016880844.1 8 - 8 4619 105
XM_017025356.1 XP_016880845.1 9 - 9 5856 105
GenBank mRNA AK023469.1 - 2 - 2 1748 105


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE164542.1 - 1 Chr. 17 - 16351217 - 16351555 [-] on Build GRCh38 105
GenBankmRNA CF595829.1 - 1 455 Chr. 17 - 16350934 - 16351675 [+] on Build GRCh38 105


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream centromere protein V CENPV 3110013H01Rik, CENP-V, PRR6, p30 Anti-sense Chr. - 16342534 - 16353498 on Build GRCh38
Downstream ubiquitin B UBB HEL-S-50 29.0kb Sense Chr. - 16380793 - 16382745 on Build GRCh38

View Full Product Details