Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC100129721
- Entrez Gene ID
-
100129721
- Gene Name
- uncharacterized LOC100129721
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK126643.1 |
- |
1 - 1 |
5346 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
family with sequence similarity 13 member C |
FAM13C |
FAM13C1 |
-340.6kb
|
Sense |
Chr. - 59245933 - 59363186 on Build GRCh38 |
Downstream |
myoregulin |
MRLN |
LINC00948, M1, MLN, MUSER1 |
27.4kb
|
Sense |
Chr. - 59736990 - 59753445 on Build GRCh38 |
View Full Product Details