Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EPOR
- Entrez Gene ID
-
2057
- Gene Name
- erythropoietin receptor
- Gene Aliases
-
EPO-R
- Chromosome Location
-
Chr.19: 11377205 - 11384342 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X97671.1 |
- |
1 - 1 |
256 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1 |
PKHD1L1 |
PKHDL1 |
|
Anti-sense |
Chr. - 109362477 - 109536809 on Build GRCh38 |
Downstream |
estrogen receptor binding site associated, antigen, 9 |
EBAG9 |
EB9, PDAF |
125.8kb
|
Anti-sense |
Chr. - 109539700 - 109565996 on Build GRCh38 |
View Full Product Details