Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLYCTK
Entrez Gene ID
132158
Gene Name
glycerate kinase
Gene Aliases
HBEBP2, HBEBP4, HBeAgBP4A
Chromosome Location
Chr.3: 52287558 - 52295256 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026702.1 3 - 4 624 85
GenBank mRNA AY172690.1 - 3 - 4 624 85


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream GLYCTK antisense RNA 1 GLYCTK-AS1 -1.2kb Anti-sense Chr. - 52288580 - 52289733 on Build GRCh38
Downstream microRNA 135a-1 MIR135A1 MIRN135-1, MIRN135A1, mir-135a-1 1.0kb Anti-sense Chr. - 52294219 - 52294308 on Build GRCh38

View Full Product Details