Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARL6IP6
- Entrez Gene ID
-
151188
- Gene Name
- ADP ribosylation factor like GTPase 6 interacting protein 6
- Gene Aliases
-
AIP-6, AIP6, PFAAP1
- Chromosome Location
-
Chr.2: 152717893 - 152761253 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024526.2 |
|
1 - 2 |
101 |
98 |
GenBank mRNA |
AW967583.1 |
- |
1 - 2 |
56 |
98 |
BP200063.1 |
- |
1 - 2 |
85 |
98 |
BX437990.2 |
- |
1 - 2 |
102 |
98 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF667868.1 |
- |
|
58 |
|
Chr. - - [-] on Build |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
formin like 2 |
FMNL2 |
FHOD2 |
-68.1kb
|
Sense |
Chr. - 152335170 - 152649834 on Build GRCh38 |
Downstream |
reprimo, TP53 dependent G2 arrest mediator candidate |
RPRM |
REPRIMO |
756.8kb
|
Anti-sense |
Chr. - 153477338 - 153478808 on Build GRCh38 |
View Full Product Details