Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IDI2-AS1
Entrez Gene ID
55853
Gene Name
IDI2 antisense RNA 1
Gene Aliases
C10orf110, HT009, IDI2-AS
Chromosome Location
Chr.10: 1022637 - 1044201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024628.1 2 - 3 305 103
NR_027708.1 3 - 4 438 103
NR_027709.1 3 - 4 522 103
GenBank mRNA BC104155.1 - 2 - 3 195 103
BC104156.1 - 2 - 3 195 103
BC112975.1 - 2 - 3 195 103
BC112976.1 - 2 - 3 195 103


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG034117.1 - 3 - 4 415 Chr. 10 - 1022660 - 1043682 [+] on Build GRCh38 103
GenBankmRNA BG334515.1 - 2 - 3 275 Chr. 10 - 1022667 - 1043900 [+] on Build GRCh38 103


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream isopentenyl-diphosphate delta isomerase 2 IDI2 IPPI2 -10.0kb Anti-sense Chr. - 1018907 - 1025859 on Build GRCh38
Downstream isopentenyl-diphosphate delta isomerase 1 IDI1 IPP1, IPPI1 3.1kb Anti-sense Chr. - 1039419 - 1056716 on Build GRCh38

View Full Product Details