The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025356.2 | NP_001020527.2 | 11 - 12 | 1643 | 74 |
NM_001142678.1 | NP_001136150.1 | 11 - 12 | 1490 | 74 | |
NM_001142679.1 | NP_001136151.1 | 11 - 12 | 1643 | 74 | |
NM_001204803.1 | NP_001191732.1 | 12 - 13 | 1706 | 74 | |
XM_005268706.4 | XP_005268763.1 | 11 - 12 | 1597 | 74 | |
XM_005268707.3 | XP_005268764.1 | 10 - 11 | 1517 | 74 | |
GenBank mRNA | AK126409.1 | - | 11 - 12 | 1724 | 74 |
AK302819.1 | - | 12 - 13 | 1500 | 74 | |
AK303996.1 | - | 11 - 12 | 1574 | 74 | |
AL832340.1 | - | 11 - 12 | 1490 | 74 | |
BC098410.1 | - | 11 - 12 | 1510 | 74 | |
BC136445.1 | - | 12 - 13 | 1399 | 74 |