The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025356.2 | NP_001020527.2 | 18 - 19 | 2752 | 63 |
NM_001142678.1 | NP_001136150.1 | 18 - 19 | 2599 | 63 | |
NM_001142679.1 | NP_001136151.1 | 18 - 19 | 2752 | 63 | |
NM_001204803.1 | NP_001191732.1 | 19 - 20 | 2815 | 63 | |
XM_005268706.4 | XP_005268763.1 | 18 - 19 | 2706 | 63 | |
XM_005268707.3 | XP_005268764.1 | 17 - 18 | 2626 | 63 | |
GenBank mRNA | AK302819.1 | - | 19 - 20 | 2609 | 63 |
AK310641.1 | - | 18 - 19 | 2719 | 63 | |
AL832340.1 | - | 18 - 19 | 2599 | 63 | |
AL833405.1 | - | 1 - 2 | 105 | 63 | |
BC098410.1 | - | 18 - 19 | 2619 | 63 | |
BC136445.1 | - | 19 - 20 | 2508 | 63 |