Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRRM5
Entrez Gene ID
100170229
Gene Name
serine/arginine repetitive matrix 5
Gene Aliases
ZNF576
Chromosome Location
Chr.19: 43612101 - 43614498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145641.1 NP_001139113.1 1 - 1 2055 73
GenBank mRNA AK093056.1 - 1 - 1 2049 73
BC034980.1 - 2 - 2 1700 73
BM679887.1 - 1 - 1 73

View Full Product Details