Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG957738.1 - 1 266 Chr. 20 - 33618761 - 33628960 [+] on Build GRCh38 63
GenBankmRNA AK021595.1 - 1 1055 Chr. 20 - 33617950 - 33619432 [+] on Build GRCh38 63


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream CBFA2/RUNX1 translocation partner 2 CBFA2T2 EHT, MTGR1, ZMYND3, p85 Sense Chr. - 33490068 - 33650031 on Build GRCh38
Downstream N-terminal EF-hand calcium binding protein 3 NECAB3 APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5 37.7kb Anti-sense Chr. - 33657087 - 33674458 on Build GRCh38

View Full Product Details