Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF088025.1 |
- |
1 |
554 |
|
Chr. 4 - 151407513 - 151408149 [+] on Build GRCh38 |
96 |
GenBankmRNA |
W15446.1 |
- |
|
|
|
Chr. - - [-] on Build |
96 |
GenBankmRNA |
AA935795.1 |
- |
1 |
|
|
Chr. 4 - 151407629 - 151408155 [-] on Build GRCh38 |
96 |
GenBankmRNA |
AA772411.1 |
- |
1 |
|
|
Chr. 4 - 151407767 - 151408155 [-] on Build GRCh38 |
96 |
GenBankmRNA |
AW663719.1 |
- |
1 |
|
|
Chr. 4 - 151407640 - 151408139 [-] on Build GRCh38 |
96 |
GenBankmRNA |
BX093436.1 |
- |
1 |
544 |
|
Chr. 4 - 151407513 - 151408146 [+] on Build GRCh38 |
96 |
GenBankmRNA |
AA988058.1 |
- |
1 |
|
|
Chr. 4 - 151407652 - 151408097 [-] on Build GRCh38 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
protease, serine 48 |
PRSS48 |
ESSPL |
-115.9kb
|
Sense |
Chr. - 151276482 - 151291856 on Build GRCh38 |
Downstream |
family with sequence similarity 160 member A1 |
FAM160A1 |
|
1.0kb
|
Sense |
Chr. - 151409143 - 151670503 on Build GRCh38 |
View Full Product Details