Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MZF1
- Entrez Gene ID
-
7593
- Gene Name
- myeloid zinc finger 1
- Gene Aliases
-
MZF-1, MZF1B, ZFP98, ZNF42, ZSCAN6
- Chromosome Location
-
Chr.19: 58561917 - 58573851 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF147345.1 |
- |
1 - 1 |
49 |
62 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
T56582.1 |
- |
|
68 |
|
Chr. - - [-] on Build |
62 |
GenBankmRNA |
DA183213.1 |
- |
1 |
108 |
|
Chr. 19 - 58571623 - 58572168 [-] on Build GRCh38 |
62 |
GenBankmRNA |
DB356944.1 |
- |
1 |
|
|
Chr. 19 - 58571696 - 58572244 [+] on Build GRCh38 |
62 |
GenBankmRNA |
AW341225.1 |
- |
1 |
|
|
Chr. 19 - 58571700 - 58572163 [+] on Build GRCh38 |
62 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MZF1 antisense RNA 1 |
MZF1-AS1 |
|
|
Anti-sense |
Chr. - 58559186 - 58574797 on Build GRCh38 |
Downstream |
CENPB DNA-binding domains containing 1 pseudogene 1 |
CENPBD1P1 |
|
3.3kb
|
Anti-sense |
Chr. - 58575399 - 58584395 on Build GRCh38 |
View Full Product Details