Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK096730.1 - 1 1751 Chr. 16 - 4813137 - 4815093 [-] on Build GRCh38 68
GenBankmRNA DB328616.1 - 1 Chr. 16 - 4813137 - 4813677 [+] on Build GRCh38 68
GenBankmRNA DW420647.1 - 1 150 Chr. 16 - 4813193 - 4813481 [-] on Build GRCh38 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream glyoxylate reductase 1 homolog GLYR1 BM045, HIBDL, N-PAC, NP60 Sense Chr. - 4803203 - 4847388 on Build GRCh38
Downstream ubinuclein 1 UBN1 VT, VT4 32.4kb Anti-sense Chr. - 4846665 - 4882401 on Build GRCh38

View Full Product Details