Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE675584.1 |
- |
1 |
|
|
Chr. 19 - 16586928 - 16587406 [+] on Build GRCh38 |
98 |
GenBankmRNA |
AU311591.1 |
- |
1 |
|
|
Chr. 19 - 16586916 - 16587494 [+] on Build GRCh38 |
98 |
GenBankmRNA |
BC030138.1 |
- |
1 |
752 |
|
Chr. 19 - 16586903 - 16587985 [-] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 35 member E1 |
SLC35E1 |
|
-14.5kb
|
Sense |
Chr. - 16549837 - 16572382 on Build GRCh38 |
Downstream |
small integral membrane protein 7 |
SMIM7 |
C19orf42 |
42.8kb
|
Sense |
Chr. - 16630741 - 16660177 on Build GRCh38 |
View Full Product Details