Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC100288619
- Entrez Gene ID
-
100288619
- Gene Name
- uncharacterized LOC100288619
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK130873.1 |
- |
1 - 2 |
33 |
60 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105376341 |
LOC105376341 |
|
-14.8kb
|
Sense |
Chr. - 951546 - 956381 on Build GRCh38 |
Downstream |
GTP binding protein 4 |
GTPBP4 |
CRFG, NGB, NOG1 |
68.0
|
Anti-sense |
Chr. - 988409 - 1017768 on Build GRCh38 |
View Full Product Details