Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC101929718
- Entrez Gene ID
-
101929718
- Gene Name
- uncharacterized LOC101929718
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 20995837 - 20999163 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110139.1 |
|
1 - 2 |
392 |
80 |
GenBank mRNA |
AK026285.1 |
- |
1 - 2 |
392 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
methyltransferase like 17 |
METTL17 |
METT11D1 |
-645.0
|
Anti-sense |
Chr. - 20989806 - 20997035 on Build GRCh38 |
Downstream |
solute carrier family 39 member 2 |
SLC39A2 |
6A1, ETI-1, ZIP-2, ZIP2 |
92.0
|
Anti-sense |
Chr. - 20999255 - 21001875 on Build GRCh38 |
View Full Product Details