Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB335958.1 - 1 Chr. 19 - 6894994 - 6895513 [+] on Build GRCh38 70
GenBankmRNA AK057344.1 - 1 1745 Chr. 19 - 6894994 - 6896832 [-] on Build GRCh38 70


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream vav guanine nucleotide exchange factor 1 VAV1 VAV -37.6kb Anti-sense Chr. - 6772668 - 6857366 on Build GRCh38
Downstream adhesion G protein-coupled receptor E4, pseudogene ADGRE4P EMR4, EMR4P, FIRE, GPR127, PGR16 55.7kb Sense Chr. - 6952500 - 6990846 on Build GRCh38

View Full Product Details