Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CD675481.1 |
- |
1 |
416 |
|
Chr. 19 - 16637351 - 16637965 [-] on Build GRCh38 |
110 |
GenBankmRNA |
AU119573.1 |
- |
1 |
557 |
|
Chr. 19 - 16637233 - 16638106 [-] on Build GRCh38 |
110 |
GenBankmRNA |
AK021856.1 |
- |
1 |
557 |
|
Chr. 19 - 16636394 - 16638106 [-] on Build GRCh38 |
110 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small integral membrane protein 7 |
SMIM7 |
C19orf42 |
|
Sense |
Chr. - 16630741 - 16660177 on Build GRCh38 |
Downstream |
transmembrane protein 38A |
TMEM38A |
TRIC-A, TRICA |
23.0kb
|
Anti-sense |
Chr. - 16661127 - 16689020 on Build GRCh38 |
View Full Product Details