Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BOLA3-AS1
Entrez Gene ID
100507171
Gene Name
BOLA3 antisense RNA 1 (head to head)
Gene Aliases
-
Chromosome Location
Chr.2: 74147981 - 74152389 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045634.1 2 - 2 3479 102
NR_045636.1 3 - 3 3353 102


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK123865.1 - 1 441 Chr. 2 - 74151238 - 74154192 [+] on Build GRCh38 102
GenBankmRNA DA774973.1 - 1 441 Chr. 2 - 74151238 - 74151799 [+] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream bolA family member 3 BOLA3 MMDS2 -3.3kb Anti-sense Chr. - 74135401 - 74147912 on Build GRCh38
Downstream MOB kinase activator 1A MOB1A C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B 800.0 Anti-sense Chr. - 74152598 - 74178967 on Build GRCh38

View Full Product Details