Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNX8
- Entrez Gene ID
-
29886
- Gene Name
- sorting nexin 8
- Gene Aliases
-
Mvp1
- Chromosome Location
-
Chr.7: 2251770 - 2354481 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
nudix hydrolase 1 |
NUDT1 |
MTH1 |
-620.0
|
Anti-sense |
Chr. - 2242222 - 2251145 on Build GRCh38 |
Downstream |
microRNA 6836 |
MIR6836 |
hsa-mir-6836 |
2.5kb
|
Sense |
Chr. - 2257515 - 2257577 on Build GRCh38 |
View Full Product Details