Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX640693.1 - 2 2755 Chr. 3 - 139678620 - 139682564 [-] on Build GRCh38 87


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream nicotinamide nucleotide adenylyltransferase 3 NMNAT3 FKSG76, PNAT-3, PNAT3 -577.0 Sense Chr. - 139560180 - 139678050 on Build GRCh38
Downstream calsyntenin 2 CLSTN2 ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma 254.2kb Anti-sense Chr. - 139935185 - 140577397 on Build GRCh38

View Full Product Details