Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK026867.1 - 1 250 Chr. 19 - 36127767 - 36129140 [+] on Build GRCh38 80


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream tubulin folding cofactor B TBCB CG22, CKAP1, CKAPI -1.8kb Sense Chr. - 36114949 - 36125948 on Build GRCh38
Downstream calpain small subunit 1 CAPNS1 CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1 10.8kb Sense Chr. - 36139926 - 36150353 on Build GRCh38

View Full Product Details