The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134774.1 | NP_001128246.1 | 9 - 10 | 1217 | 87 |
NM_001134775.1 | NP_001128247.1 | 10 - 11 | 1511 | 87 | |
NM_001134776.1 | NP_001128248.1 | 10 - 11 | 1390 | 87 | |
NM_001318734.1 | NP_001305663.1 | 10 - 11 | 1448 | 87 | |
NM_022822.2 | NP_073733.1 | 10 - 11 | 1485 | 87 | |
XM_005274183.1 | XP_005274240.1 | 10 - 11 | 1427 | 87 | |
XM_005274184.1 | XP_005274241.1 | 10 - 11 | 1448 | 87 | |
XM_017018133.1 | XP_016873622.1 | 8 - 9 | 1384 | 87 | |
GenBank mRNA | AK022449.1 | - | 10 - 11 | 1377 | 87 |
AK022907.1 | - | 10 - 11 | 1430 | 87 | |
AK094593.1 | - | 8 - 9 | 1087 | 87 | |
AK126489.1 | - | 8 - 9 | 1869 | 87 | |
AK315725.1 | - | 10 - 11 | 1380 | 87 | |
AL136864.1 | - | 10 - 11 | 1411 | 87 | |
BC034373.1 | - | 10 - 11 | 1373 | 87 |