Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR6N2
Entrez Gene ID
81442
Gene Name
olfactory receptor family 6 subfamily N member 2
Gene Aliases
OR1-23
Chromosome Location
Chr.1: 158776682 - 158777635 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005278.1 NP_001005278.1 1 - 1 653 97

View Full Product Details