Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAP2
Entrez Gene ID
55620
Gene Name
signal transducing adaptor family member 2
Gene Aliases
BKS
Chromosome Location
Chr.19: 4324043 - 4338877 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013841.1 NP_001013863.1 5 - 6 550 82
NM_017720.2 NP_060190.2 5 - 6 550 82
XM_011528123.1 XP_011526425.1 5 - 6 577 82
GenBank mRNA AJ245719.1 - 5 - 6 511 82
AK000241.1 - 5 - 6 521 82
AK125774.1 - 7 - 8 1060 82
BC000795.1 - 5 - 6 501 82

View Full Product Details