The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025071.1 | NP_001020242.1 | 1 - 2 | 278 | 97 |
GenBank mRNA | BM557014.1 | - | 1 - 2 | 213 | 97 |