Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIX6
Entrez Gene ID
4990
Gene Name
SIX homeobox 6
Gene Aliases
MCOPCT2, ODRMD, OPTX2, Six9
Chromosome Location
Chr.14: 60509220 - 60511807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007374.2 NP_031400.2 1 - 2 749 94
GenBank mRNA AJ011785.1 - 1 - 2 644 94
BC065831.1 - 1 - 2 749 94
BC069413.1 - 1 - 2 653 94
BC087852.1 - 1 - 2 727 94

View Full Product Details