Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSD
Entrez Gene ID
414
Gene Name
arylsulfatase D
Gene Aliases
ASD
Chromosome Location
Chr.X: 2903970 - 2929375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001669.3 NP_001660.2 2 - 3 294 104
NM_009589.3 NP_033667.2 2 - 3 294 104
XM_005274514.2 XP_005274571.1 2 - 3 264 104
XM_005274515.2 XP_005274572.1 2 - 3 264 104
GenBank mRNA AF160499.1 - 2 - 3 194 104
AK314479.1 - 2 - 3 258 104
BC020229.1 - - 222 104
BC080179.1 - 2 - 3 197 104
DA869167.1 - 2 - 3 294 104
X83572.1 - 2 - 3 270 104

View Full Product Details