Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC43
Entrez Gene ID
124808
Gene Name
coiled-coil domain containing 43
Gene Aliases
-
Chromosome Location
Chr.17: 44677437 - 44689797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144609.2 NP_653210.2 4 - 5 522 86
GenBank mRNA BC023298.1 - 4 - 5 483 86
BC047776.2 - 4 - 5 497 86

View Full Product Details