Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFP62
Entrez Gene ID
643836
Gene Name
ZFP62 zinc finger protein
Gene Aliases
ZET, ZNF755
Chromosome Location
Chr.5: 180843580 - 180861327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152283.4 NP_689496.4 2 - 3 137 66
XM_017009712.1 XP_016865201.1 3 - 4 240 66
XM_017009713.1 XP_016865202.1 3 - 4 288 66
XM_017009714.1 XP_016865203.1 2 - 3 221 66
XM_017009721.1 XP_016865210.1 2 - 3 147 66
GenBank mRNA AK091376.1 - 2 - 3 831 66
AK295714.1 - 2 - 3 127 66
BC029573.1 - 2 - 3 136 66
DA216419.1 - 2 - 3 110 66
DA323041.1 - 2 - 3 127 66

View Full Product Details