Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRIM64
- Entrez Gene ID
-
120146
- Gene Name
- tripartite motif containing 64
- Gene Aliases
-
C11orf28, TRIM64A
- Chromosome Location
-
Chr.11: 89966068 - 89974147 on Build GRCh38
- Gene Symbol
- TRIM64B
- Entrez Gene ID
-
642446
- Gene Name
- tripartite motif containing 64B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 89870188 - 89878456 on Build GRCh38
- Gene Symbol
- TRIM64EP
- Entrez Gene ID
-
440061
- Gene Name
- tripartite motif containing 64E, pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 90057419 - 90062997 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY010113.1 |
- |
4 - 5 |
490 |
96 |
View Full Product Details