Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D3F
Entrez Gene ID
84218
Gene Name
TBC1 domain family member 3F
Gene Aliases
TBC1D3
Chromosome Location
Chr.17: 36428612 - 36439523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032258.4 NP_115634.3 13 - 14 1202 103
GenBank mRNA AK225985.1 - 13 - 14 1193 103

Gene Symbol
LOC101060389
Entrez Gene ID
101060389
Gene Name
TBC1 domain family member-like
Gene Aliases
TBC1D3D
Chromosome Location
Chr.17: 38004033 - 38014908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291465.1 NP_001278394.1 13 - 14 1188 103
XM_006722248.2 XP_006722311.1 12 - 13 2694 103
XM_006722249.2 XP_006722312.1 11 - 12 2628 103
XM_006722252.3 XP_006722315.1 11 - 12 2445 103
XM_011524176.2 XP_011522478.1 13 - 14 1301 103

Gene Symbol
TBC1D3
Entrez Gene ID
729873
Gene Name
TBC1 domain family member 3
Gene Aliases
PRC17, TBC1D3A, TBC1D3F
Chromosome Location
Chr.17: 38181476 - 38192555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123391.3 NP_001116863.3 13 - 14 1200 103
GenBank mRNA AB449884.1 - 12 - 13 1088 103
AK302064.1 - 14 - 15 1426 103
AK303259.1 - 12 - 13 1220 103
AX775941.1 - 15 - 16 1449 103
AX775943.1 - 15 - 16 1449 103

Gene Symbol
TBC1D3B
Entrez Gene ID
414059
Gene Name
TBC1 domain family member 3B
Gene Aliases
PRC17, TBC1D3I, TBC1D3L
Chromosome Location
Chr.17: 36165681 - 36176624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001417.6 NP_001001417.6 13 - 14 1234 103
XM_005257980.4 XP_005258037.2 13 - 14 1364 103
GenBank mRNA AF540953.1 - 12 - 13 1088 103
AK292257.1 - 13 - 14 1205 103

Gene Symbol
TBC1D3C
Entrez Gene ID
414060
Gene Name
TBC1 domain family member 3C
Gene Aliases
TBC1D3D
Chromosome Location
Chr.17: 38057693 - 38068634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001418.6 NP_001001418.5 13 - 14 1228 103
XM_006721904.2 XP_006721967.1 12 - 13 2694 103
XM_006721905.2 XP_006721968.1 11 - 12 2628 103
XM_006721907.3 XP_006721970.1 11 - 12 2445 103
XM_011524814.2 XP_011523116.1 13 - 14 1301 103
GenBank mRNA AA045947.1 - 1 - 2 116 103
AK313329.1 - 13 - 14 1228 103

Gene Symbol
TBC1D3E
Entrez Gene ID
102723859
Gene Name
TBC1 domain family member 3E
Gene Aliases
TBC1DE3
Chromosome Location
Chr.17: 38119293 - 38138868 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291466.1 NP_001278395.1 13 - 14 1186 103
XM_006722254.1 XP_006722317.1 16 - 17 1535 103
GenBank mRNA AL136860.1 - 13 - 14 1186 103
BC047739.1 - 14 - 15 1592 103

Gene Symbol
TBC1D3G
Entrez Gene ID
101060321
Gene Name
TBC1 domain family member 3G
Gene Aliases
-
Chromosome Location
Chr.17: 36322242 - 36334759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291462.1 NP_001278391.1 13 - 14 1188 103
XM_005276914.3 XP_005276971.1 13 - 14 3013 103
GenBank mRNA AK292308.1 - 14 - 15 1238 103
AK301823.1 - 11 - 12 1000 103
BC071680.1 - 13 - 14 1188 103

Gene Symbol
TBC1D3H
Entrez Gene ID
729877
Gene Name
TBC1 domain family member 3H
Gene Aliases
-
Chromosome Location
Chr.17: 36377475 - 36388448 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123392.3 NP_001116864.3 13 - 14 1202 103
XM_006722055.1 XP_006722118.1 12 - 13 1842 103
XM_006722056.1 XP_006722119.1 11 - 12 1776 103
XM_011525172.1 XP_011523474.1 12 - 13 2025 103
XM_011525173.2 XP_011523475.1 13 - 14 1396 103
XM_011525174.1 XP_011523476.1 11 - 12 1959 103
GenBank mRNA EL953327.1 - 2 - 3 191 103

Gene Symbol
TBC1D3I
Entrez Gene ID
102724862
Gene Name
TBC1 domain family member 3I
Gene Aliases
-
Chromosome Location
Chr.17: 36253639 - 36264601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291463.1 NP_001278392.1 13 - 14 1188 103
XM_006722223.3 XP_006722286.1 13 - 14 1459 103
XM_006722224.3 XP_006722287.1 12 - 13 2704 103
XM_006722226.3 XP_006722289.1 11 - 12 2638 103
XM_011524183.2 XP_011522485.1 11 - 12 2821 103
GenBank mRNA BC078140.1 - 11 - 12 1023 103

Gene Symbol
TBC1D3K
Entrez Gene ID
101060351
Gene Name
TBC1 domain family member 3K
Gene Aliases
-
Chromosome Location
Chr.17: 37923805 - 37935371 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291464.1 NP_001278393.1 13 - 14 1188 103
XM_006722235.2 XP_006722298.1 12 - 13 3466 103
XM_006722236.1 XP_006722299.1 13 - 14 2038 103
XM_006722237.2 XP_006722300.1 12 - 13 3283 103
XM_006722239.2 XP_006722302.1 11 - 12 3217 103
XM_011524172.1 XP_011522474.1 11 - 12 3400 103

Gene Symbol
TBC1D3L
Entrez Gene ID
101060376
Gene Name
TBC1 domain family member 3L
Gene Aliases
-
Chromosome Location
Chr.17: 37978155 - 37990691 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291459.1 NP_001278388.1 13 - 14 1202 103
XM_006722242.3 XP_006722305.1 12 - 13 4442 103
XM_006722243.2 XP_006722306.1 11 - 12 2628 103
XM_006722246.3 XP_006722309.1 11 - 12 1776 103
XM_011524175.2 XP_011522477.1 13 - 14 1301 103
GenBank mRNA AK095385.1 - 11 - 12 1776 103
AK122833.1 - 9 - 10 2513 103
AX775949.1 - 11 - 12 1776 103
AX775951.1 - 11 - 12 1776 103

View Full Product Details