Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
NBPF15
Entrez Gene ID
284565
Gene Name
neuroblastoma breakpoint family member 15
Gene Aliases
AB14, AG3, NBPF16
Chromosome Location
Chr.1: 144421386 - 144459464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170755.2 NP_001164226.1 5 - 5 553 75
GenBank mRNA AK097180.1 - 5 - 5 472 75

View Full Product Details