The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000677.3 | NP_000668.1 | 2 - 2 | 1454 | 97 |
NM_001302678.1 | NP_001289607.1 | 2 - 2 | 1518 | 97 | |
NM_001302679.1 | NP_001289608.1 | 2 - 2 | 960 | 97 | |
GenBank mRNA | AK313095.1 | - | 2 - 2 | 1048 | 97 |
AY136749.1 | - | 2 - 2 | 687 | 97 | |
CD013875.1 | - | 2 - 2 | 809 | 97 | |
JN855714.1 | - | 2 - 2 | 686 | 97 | |
L20463.1 | - | 2 - 2 | 978 | 97 | |
L22607.1 | - | 2 - 2 | 952 | 97 | |
X76981.1 | - | 2 - 2 | 1020 | 97 |