Product Details

Assay Reference Genome
Location

Chr.1:196985627 on build GRCh38
Cytoband
1q31.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
81494
Gene Symbol
CFHR5
Gene Name
complement factor H related 5
Gene Aliases
CFHL5, CFHR5D, FHR-5, FHR5
Location
Chr.1:196975022-197009725 on build GRCh38
Assay Gene Location
Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CFHR5 NM_030787.3 NP_110414.1
XM_011510020.2 XP_011508322.1
AF295327.1 AAK15619.1
AK314026.1
BC026282.1
BC111773.1 AAI11774.1
DA638244.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv12e55 Chr1:196762018 - 197042447 on Build GRCh38 Gain CFHR4 CFHR1 F13B CFHR5 CFHR3 CFHR2
nsv469599 Chr1:196827346 - 196986100 on Build GRCh38 Loss CFHR4 CFHR1 CFHR5 CFHR2
nsv428290 Chr1:196621707 - 196992672 on Build GRCh38 Gain+Loss CFHR4 CFHR1 CFHR5 CFH CFHR3 CFHR2
nsv471614 Chr1:196700381 - 196986100 on Build GRCh38 Gain+Loss CFHR4 CFHR1 CFHR5 CFH CFHR3 CFHR2
nsv528460 Chr1:196573437 - 197477572 on Build GRCh38 Gain MIR4735 CFHR4 CFHR1 F13B CFHR5 ZBTB41 ASPM CFH CFHR3 KCNT2 CRB1 CFHR2

View Full Product Details