Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC100303749
- Entrez Gene ID
-
100303749
- Gene Name
- LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae) pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 65944805 - 65945573 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_028474.1 |
|
1 - 1 |
154 |
117 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU568479.1 |
- |
|
318 |
|
Chr. - - [-] on Build |
117 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
neurolysin |
NLN |
AGTBP, EP24.16, MEP, MOP |
-115.5kb
|
Anti-sense |
Chr. - 65722196 - 65829283 on Build GRCh38 |
Downstream |
splicing regulatory glutamic acid and lysine rich protein 1 |
SREK1 |
SFRS12, SRrp508, SRrp86 |
198.6kb
|
Anti-sense |
Chr. - 66144218 - 66183616 on Build GRCh38 |
View Full Product Details