Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKN2B-AS1
Entrez Gene ID
100048912
Gene Name
CDKN2B antisense RNA 1
Gene Aliases
ANRIL, CDKN2B-AS, CDKN2BAS, NCRNA00089, PCAT12, p15AS
Chromosome Location
Chr.9: 21994791 - 22121097 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003529.3 16 - 17 2910 132
NR_047532.1 11 - 12 1699 132
NR_047534.1 6 - 7 963 132
NR_047543.1 6 - 7 992 132
GenBank mRNA AB548314.1 - 11 - 12 1699 132
DQ485453.1 - 16 - 17 2909 132
GQ495923.1 - 6 - 7 881 132
GQ495925.1 - 6 - 7 963 132


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX100299.1 - 3 - 4 264 Chr. 9 - 22097313 - 22120321 [+] on Build GRCh38 132


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream cyclin dependent kinase inhibitor 2B CDKN2B CDK4I, INK4B, MTS2, P15, TP15, p15INK4b -104.4kb Anti-sense Chr. - 22002903 - 22009313 on Build GRCh38
Downstream DMRT like family A1 DMRTA1 DMO, DMRT4 328.1kb Sense Chr. - 22446841 - 22452473 on Build GRCh38

View Full Product Details