Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1orf54
Entrez Gene ID
79630
Gene Name
chromosome 1 open reading frame 54
Gene Aliases
-
Chromosome Location
Chr.1: 150272284 - 150280927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301040.1 NP_001287969.1 5 - 6 451 105
NM_024579.3 NP_078855.2 4 - 5 340 105
GenBank mRNA AI004135.1 - 2 - 3 105
AK026874.1 - 4 - 5 325 105
AX770997.1 - 4 - 5 338 105

View Full Product Details