The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301852.1 | NP_001288781.1 | 5 - 6 | 494 | 111 |
XM_011532896.2 | XP_011531198.1 | 4 - 5 | 387 | 111 |