The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297575.1 | NP_001284504.1 | 5 - 6 | 683 | 102 |
NM_014625.3 | NP_055440.1 | 6 - 7 | 887 | 102 | |
XM_005245483.3 | XP_005245540.1 | 4 - 5 | 732 | 102 | |
GenBank mRNA | AJ279254.1 | - | 6 - 7 | 868 | 102 |
BC029141.1 | - | 5 - 6 | 683 | 102 |