The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294345.1 | NP_001281274.1 | 2 - 3 | 297 | 109 |
NM_001294346.1 | NP_001281275.1 | 2 - 3 | 404 | 109 | |
NM_016533.5 | NP_057617.2 | 2 - 3 | 643 | 109 | |
GenBank mRNA | AF205633.1 | - | 2 - 3 | 318 | 109 |
BC057766.1 | - | 2 - 3 | 311 | 109 | |
BI597990.1 | - | 2 - 3 | 496 | 109 | |
BM128205.1 | - | 2 - 3 | 265 | 109 | |
CB995632.1 | - | 2 - 3 | 324 | 109 | |
CD695373.1 | - | 2 - 3 | 391 | 109 |