Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGS1
Entrez Gene ID
9489
Gene Name
phosphatidylglycerophosphate synthase 1
Gene Aliases
-
Chromosome Location
Chr.17: 78378618 - 78424659 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_111989.1 1 - 2 192 83
GenBank mRNA AK024529.1 - 1 - 2 161 83
AK225030.1 - 1 - 2 161 83


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BP381508.1 - 1 - 2 161 Chr. 17 - 78378649 - 78400827 [+] on Build GRCh38 83


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101928674 LOC101928674 -4.7kb Sense Chr. - 78360448 - 78373909 on Build GRCh38
Downstream dynein axonemal heavy chain 17 DNAH17 DNAHL1, DNEL2 27.3kb Anti-sense Chr. - 78423697 - 78577430 on Build GRCh38

View Full Product Details