Product Details
- Assay Reference Genome
Location
Chr.2:166231086 on build GRCh38
- Cytoband
- 2q24.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 101929680
- Gene Symbol
- LOC101929680
- Gene Name
- uncharacterized LOC101929680
- Gene Aliases
- Location
- Chr.2:166081531-166301784 on build GRCh38
- Assay Gene Location
- Within Intron 4
- Entrez Gene ID
- 6335
- Gene Symbol
- SCN9A
- Gene Name
- sodium voltage-gated channel alpha subunit 9
- Gene Aliases
- ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
- Location
- Chr.2:166195185-166375987 on build GRCh38
- Assay Gene Location
- Within Intron 23
View Full Product Details