Product Details

Assay Reference Genome
Location

Chr.15:66397465 on build GRCh38
Cytoband
15q22.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5604
Gene Symbol
MAP2K1
Gene Name
mitogen-activated protein kinase kinase 1
Gene Aliases
CFC3, MAPKK1, MEK1, MKK1, PRKMK1
Location
Chr.15:66386873-66491544 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
MAP2K1 NM_002755.3 NP_002746.1
XM_017022411.1 XP_016877900.1
AK291500.1
AK312356.1
BC137459.1
BC139729.1
L05624.1 AAA36318.1
L11284.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833045 Chr15:66295248 - 66460419 on Build GRCh38 Gain TIPIN SCARNA14 DIS3L MAP2K1

View Full Product Details