Gene Transcripts

Gene Symbol
Fbxo4
Entrez Gene ID
106052
Gene Name
F-box protein 4
Gene Aliases
1700096C12Rik, AI851261, AW494535, Fbx4
Chromosome Location
Chr. 15: 3963564 - 3979573 on Build GRCm38
UniGene ID
Mm.234191
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134099.2 NP_598860.2 2 502
GenBank mRNA BC036385.1 - 2 468
BC040086.1 - 2 502 3495299

View Full Product Details