Gene Transcripts

Gene Symbol
Dnaaf2
Entrez Gene ID
109065
Gene Name
dynein, axonemal assembly factor 2
Gene Aliases
1110034A24Rik, 2810020C19Rik, AV032410, Ktu, TU54, kintoun, mknt
Chromosome Location
Chr. 12: 69189838 - 69198429 on Build GRCm38
UniGene ID
Mm.107180
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027269.3 NP_081545.3 3 2098
GenBank mRNA AB455811.1 - 3 1954
AK012767.1 - 3 776
AK075648.1 - 750
AK154254.1 - 3 803
AK172494.1 - 3 774
BC055807.1 - 3 2098 6414041
BC058344.1 - 683
BC062898.1 - 3 2098 6414041

View Full Product Details