Gene Transcripts
- Gene Symbol
- Rnf20
- Entrez Gene ID
- 109331
- Gene Name
- ring finger protein 20
- Gene Aliases
- 4833430L21Rik, AW540162, C79397, mKIAA4116
- Chromosome Location
- Chr. 4: 49632011 - 49656886 on Build GRCm38
- UniGene ID
- Mm.24765
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163263.1 |
NP_001156735.1
|
5, 6 |
1099 |
|
NM_182999.2 |
NP_892044.1
|
6, 7 |
946 |
|
GenBank mRNA |
AK048862.1 |
- |
5, 6 |
1058 |
|
AK051278.1 |
- |
5, 6 |
1058 |
|
AK053783.1 |
- |
6, 7 |
934 |
|
AK076501.1 |
- |
1, 2 |
28 |
|
AK082529.1 |
- |
5, 6 |
1003 |
|
AK163905.1 |
- |
2, 3 |
312 |
|
AK220492.1 |
- |
5, 6 |
1015 |
|
BC004593.1 |
- |
1, 2 |
173 |
|
BC052482.1 |
- |
6, 7 |
885 |
6308280 |
View Full Product Details