Gene Transcripts

Gene Symbol
Aldoc
Entrez Gene ID
11676
Gene Name
aldolase C, fructose-bisphosphate
Gene Aliases
AI847350, AU040929, Aldo3, Scrg2
Chromosome Location
Chr. 11: 78323073 - 78327781 on Build GRCm38
UniGene ID
Mm.7729
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK005077.1 - 6 924
AK039267.1 - 5 584
BC004802.1 - 7 755 3589325
BC008184.1 - 7 755 3589205
S72537.1 - 7 779

View Full Product Details