Gene Transcripts

Gene Symbol
Atf2
Entrez Gene ID
11909
Gene Name
activating transcription factor 2
Gene Aliases
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP
Chromosome Location
Chr. 2: 73816509 - 73892639 on Build GRCm38
UniGene ID
Mm.209903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025093.2 NP_001020264.1 15 1831
NM_001284369.1 NP_001271298.1 13 1671
NM_001284372.1 NP_001271301.1 13 1537
NM_001284373.1 NP_001271302.1 12 1474
NM_001284374.1 NP_001271303.1 12 1556
NM_001284376.1 NP_001271305.1 15 1713
NM_009715.3 NP_033845.1 14 1734
GenBank mRNA AF483482.1 - 11 1408
AF483483.1 - 11 1408
AK046184.1 - 4 592
AK051591.1 - 14 1724
AK139515.1 - 13 1587
AK144977.1 - 13 1511
AK145887.1 - 13 1441
AY902311.1 - 10 1294
BC042210.1 - 12 1507 4018323
BC079883.1 - 14 1656 6839572
BC082596.1 - 12 1420 30649207
BC085137.1 - 12 1499
M31629.1 - 7 886
M77167.1 - 9 1180
S76657.1 - 1499
S76659.1 - 1205
U46026.1 - 4 676

View Full Product Details